Extech Differential Pressure Manometer wth Data Logging
Records continuous data on SD card for future analysis
  • Select from 10 different units of measure
  • Store up to 99 readings manually
  • Easily analyze data with Min/Max recall function
  • Save battery life with auto power-off function
The SDL700 series differential pressure manometers/data loggers feature up to 10 types of pressure readings displayed on a large backlit LCD. Three distinct models are available — low range (2.9 psi), mid-range (29 psi), and high range (101.5 psi). Each model features dual-differential inputs with internal sensors, data Hold, and a zero function for offset correction or measurement.
Up to 99 readings can be manually stored. A SD memory card allows for continuous data logging, which is date- and time-stamped and can be easily exported to your PC as an Excel® file for further analysis.
114,000.00 - ₹117,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Extech SDL710 Differential Pressure Manometer Data Logger, 2.9 psi 4250002 Mfr # SDL710
COMPARE
±2% 2.9 0.001 psi
₹117,000.00INR / Each
Extech SDL720 Differential Pressure Manometer Data Logger, 29 psi 4250003 Mfr # SDL720
COMPARE
±2% 29 0.01 psi
₹114,000.00INR / Each
Extech SDL730 Differential Pressure Manometer Data Logger, 101.5 psi 4250004 Mfr # SDL730
COMPARE
±2% 101.5 0.05 psi
₹114,000.00INR / Each