Extech Compact Multimeters with Voltage Detector
Quickly check for the presence of AC voltage before testing
These compact multimeters feature a built-in noncontact AC voltage detector with a red LED indicator and audible beeper ensuring operator safety. Meters provide 100 to 600 VAC measurement capability with 0.5% basic accuracy. Additional features include low-battery indicator and an easy-to-read large 1" LCD digit display.
Model 20005-28 provides manual ranging capabilities with a 2000-count display. Model 20005-29 features auto-ranging capabilities with a 2000-count display. Meter includes max hold, data hold, and auto power off conserving battery life. Model 20005-30 features auto-ranging capabilities with a 4000-count display and type K thermocouple measurements (°F/°C switchable). Additional functions include capacitance, frequency, and relative function.
11,400.00 - ₹19,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) Resistance Max (MΩ)
Availability Pricing
Extech EX310 Mini Manual Ranging Multimeter + Voltage Detector 2000528 Mfr # EX310
COMPARE
600 600 2
₹11,400.00INR / Each
Extech EX330 Mini Autoranging MultiMeter with Temp + Voltage Detector 2000530 Mfr # EX330
COMPARE
600 600 40
₹19,600.00INR / Each