Extech® Big Digit Indoor/Outdoor Thermometer
Easily view minimum/maximum temperatures on large LCD
  • Programmable high/low alarms provide outdoor temperature alerts
  • Water resistant/splash proof probe
  • Internal memory stores min/max readings
  • Selectable temperature scale (°F/°C)
The large LCD indoor and outdoor thermometer features a dual display with 1” digits, plus a user programmable high and low alarm for outdoor temperature. A built-in sensor provides indoor temperature readings from 14 to 140°F (-10 to 60°C). An external probe records remote temperature from -58 to 158°F (-50 to 70°C).
The water resistant/splash proof probe with a 9.6 ft (3m) thin cable, can easy be installed through windows, doors, or any small openings leading outdoors. It is also ideal for use inside refrigerators and freezers or temperature-controlled areas. Other features include an internal memory that stores min/max readings, low-battery indicator, built-in stand for desktop display, and mounting bracket for wall display.
10,500.00 - ₹11,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Extech 401014A Big Digit In/Outdoor Thermometer with Alarm 3572508 Mfr # 401014A
COMPARE
LCD 158 -58
₹11,600.00INR / Each
Extech 401014 Big Digital In/Outdoor Thermometer 8648724 Mfr # 401014
COMPARE
₹10,500.00INR / Each