Explosion-Proof Pressure Switches
The preferred choice for plant instrumentation upgrades
  • Digital accuracy amd reliability
  • Large retatable process display
  • Programmable set point and deadband
  • Worldwide approvals for Divisions 1 and 2
  • Replaces switch, gauge, and transmitter
  • Configurable self diagnostics
  • Choose from 2-wire, loop-powered, and external powered models
Models 68093-12 to -17 have a 4 to 20 mA output and one 140 VAC/VDC switch. Models 68093-24 to -29 have 4 to 20 mA ouput and two 140 VAC/VDC switches. All other models have solid state switches.
377,000.00 - ₹517,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
United Electric 2X2D00P14 Explosion Proof Pressure Switch, 2-wire/12-30 VDC/Div 1,2/0-100psi 6809302 Mfr # 1XSWLLP14
COMPARE
±0.5% full-scale 100
₹377,000.00INR / Each
United Electric 2X2D00P16 Explosion Proof Pressure Switch, 2-wire/12-30 VDC/Div 1,2/0-500psi 6809304 Mfr # 1XSWLLP16
COMPARE
±0.5% full-scale 500
₹377,000.00INR / Each
United Electric 2W3A00P14 Explosion Proof Pressure Switch, 2-wire/90-130VDC/Div 2/0-100psi 6809309 Mfr # 1XSWHLP14
COMPARE
±0.5% full-scale 100
₹418,000.00INR / Each
United Electric 2XLP41P14 EX/ATEX 1, 2 Pressure Switch, 0-100 psi, 4-20 mA, Loop 6809314 Mfr # 1XTXSWP14
COMPARE
±0.5% full-scale 100
₹511,000.00INR / Each
United Electric 8X2D45P14 EX/ATEX 1, 2 Pressure Switch, 0-100 psi, 4-20 mA, 10-30VDC 6809326 Mfr # 1XTXSWP14
COMPARE
±0.5% full-scale 100
₹511,000.00INR / Each
United Electric 2W2D00K12 EX/ATEX 2 Differential Pressure Switch, 100 psi, DC, 40 mA 6809331 Mfr # 1XSWLLK12
COMPARE
±0.5% full-scale 100
₹517,000.00INR / Each