Excelon Micro-Line Translucent Mini-Bore Tubing
A translucent, odorless, and tasteless mini-bore tubing that offers practical solutions to maintaining a non-toxic and non-contaminating environment
  • Meets USP Class VI, FDA Regulation 21CFR177.350, and National Formulary Vo. 14
  • No plasticizers used in the manufacturing process
  • Excellent flexibility with no kinking and 100% plastic memory
  • Retains flexibility in sub-zero temperatures and resists age hardening
  • Won’t leach or migrate
This translucent mini-bore tubing provides practical solutions for challenges in biological and clinical laboratories. It is easily fabricated and can be adapted for connectors, needles, or fittings through processes such as heat-expanding, heat-shrink-joining, expanding, force fitting, threading, restoring, heat-connecting, fabricating, bending, reduce-tapering, and cutting.
13,900.00 - ₹22,400.00INR / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Excelon Micro-Line Tubing, 0.04" ID x 0.07" OD; 50 Ft 5011748 Mfr # 18160-50FT
COMPARE
1/25 5/64 1/64
₹15,100.00INR / Pkg of 1
Excelon Micro-Line Tubing, 0.05" ID x 0.09" OD; 50 Ft 5011749 Mfr # 18200-50FT
COMPARE
1/20 3/32 1/50
₹15,500.00INR / Pkg of 1
Excelon Micro-Line Tubing, 0.02" ID x 0.06" OD; 50 Ft 5011750 Mfr # 1850-50FT
COMPARE
1/50 1/16 1/50
₹13,900.00INR / Pkg of 1
Excelon Micro-Line Tubing, 0.03" ID x 0.09" OD; 50 Ft 5011751 Mfr # 1860-50FT
COMPARE
1/32 3/32 1/32
₹14,400.00INR / Pkg of 1
Excelon Micro-Line Tubing, 0.063" ID x 0.125" OD; 50 Ft 5011752 Mfr # 1861-50FT
COMPARE
1/16 1/8 1/32
₹22,400.00INR / Pkg of 1