Excelon Bev-A-Line® V HT Tubing
Adds no taste to water or beverages
  • An equivalent to Vendflow® E-70-V-CE
  • Inner and out layers are plasticizer-free
  • High-clarity, lightweight, and flexible while remaining strong and durable
  • Meets FDA requirements for food contact
  • Irradiated version of Bev-A-Line® IV
Bev-A-Line® V HT plastic tubing has a smooth inner surface that adds no taste to water and beverages. It is safe for food, beverage, pharmaceutical, laboratory, and general usage. It meets FDA, USDA, National Formulary, and USP standards. This tubing is designed to be flexible across a broad temperature range, allowing for simple installation. It is also strong and durable to reduce replacement costs.
9,200.00 - ₹161,000.00INR / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Excelon Bev-A-Line® V HT Tubing, 1/16" ID x 3/16" OD; 50 Ft 5011761 Mfr # 2150105
COMPARE
1/16 3/16 1/16
₹15,300.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 1/8" ID x 3/16" OD; 50 Ft 5011762 Mfr # 2150405
COMPARE
1/8 3/16 1/32
₹9,200.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 1/8" ID x 1/4" OD; 50 Ft 5011763 Mfr # 2150505
COMPARE
1/8 1/4 1/16
₹19,400.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 3/16" ID x 1/4" OD; 50 Ft 5011764 Mfr # 2150805
COMPARE
3/16 1/4 1/32
₹12,900.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 3/16" ID x 5/16" OD; 50 Ft 5011765 Mfr # 2150905
COMPARE
3/16 5/16 1/16
₹23,500.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 1/4" ID x 3/8" OD; 50 Ft 5011766 Mfr # 2151305
COMPARE
1/4 3/8 1/16
₹32,200.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 5/16" ID x 7/16" OD; 50 Ft 5011767 Mfr # 2151605
COMPARE
5/16 7/16 1/16
₹38,000.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 3/8" ID x 1/2" OD; 50 Ft 5011768 Mfr # 2151905
COMPARE
3/8 1/2 1/16
₹43,300.00INR / Pkg of 1
Excelon Bev-A-Line® V HT Tubing, 3/4" ID x 1" OD; 50 Ft 5011769 Mfr # 2153805
COMPARE
3/4 1 1/8
₹161,000.00INR / Pkg of 1
Showing 1- 9 of 9