Excelon Bev-A-Line® IV Tubing
A lightweight, high clarity, transparent tubing, which provides higher-pressure and vacuum capabilities
  • Meets FDA, USDA, National Formulary, and USP standards for food, beverage, pharmaceutical, and laboratory usage
  • Co-extruded to combine the benefits of two tubes in one product
  • The clear shell provides excellent durability and greater burst strength than PVC
  • Complete visual flow
  • Free technical application support available to assist with tubing selection
Bev-A-Line® IV’s low friction coefficient and great chemical resistance allow easy passage of solids, alcohols, acids, caustics, and solvents. The smooth interior surface is exceptionally resistant to the absorption of liquids, providing a neutral and stain-free environment. Its exceptional flexibility provides superior resistance to freezing and thermal cycling, without the use of vinyl chloride monomer and plasticizer.
2,900.00 - ₹74,400.00INR / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Excelon Bev-A-Line® IV Tubing, 1/8" ID x 3/16" OD; 50 Ft 5011753 Mfr # 2140405
COMPARE
1/8 3/16 1/32
₹2,900.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 1/8" ID x 1/4" OD; 50 Ft 5011754 Mfr # 2140505
COMPARE
1/8 1/4 1/16
₹6,400.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 1/4" ID x 3/8" OD; 50 Ft 5011755 Mfr # 2141305
COMPARE
1/4 3/8 1/16
₹11,100.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 3/8" ID x 1/2" OD; 50 Ft 5011756 Mfr # 2141905
COMPARE
3/8 1/2 1/16
₹15,900.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 1/2" ID x 5/8" OD; 50 Ft 5011757 Mfr # 2142505
COMPARE
1/2 5/8 1/16
₹19,900.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 1/2" ID x 3/4" OD; 50 Ft 5011758 Mfr # 2142705
COMPARE
1/2 3/4 1/8
₹42,000.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 3/4" ID x 1" OD; 50 Ft 5011759 Mfr # 2143805
COMPARE
3/4 1 1/8
₹56,000.00INR / Pkg of 1
Excelon Bev-A-Line® IV Tubing, 1" ID x 1-1/4" OD; 50 Ft 5011760 Mfr # 2144505
COMPARE
1 1 1/4 1/8
₹74,400.00INR / Pkg of 1
Showing 1- 8 of 8