Etatron DLX Series Wall-Mount pH/ORP Pump Control Systems
Integrated controller and metering pump simplify system setup
  • Control systems are able to accept pH or ORP electrodes for added convenience
  • On/off or proportional control modes allow for robust process control
Control the pH or ORP levels of your process with an integrated controller and pump—saves you time, money, and space. These solenoid-driven, diaphragm metering pumps and controllers are encased with an IP65, acid-resistant plastic housing. Polypropylene pump heads have manual air bleed, PTFE diaphragm, and Viton wetted parts. Pump control systems accept level switches (order separately). Backlit LCD display has resolution of 0.01 pH or 1 mV.
Controllers are programmable for on/off or proportional control applications and have manual temperature compensation. Control can be fine-tuned by programmable hysteresis and delay functions. Frequency of injections is programmable between 0 to 100% of the maximum flow rate of the pump.
Control systems accept electrodes with BNC connectors and no temperature compensation; order electrodes separately.
See instruction manual for complete specifications
200,000.00 - ₹246,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Pressure (PSI) Power (VAC)
Availability Pricing
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 1 L/hr, 15 bar, Wall-Mt; 115V 5602240 Mfr # PLX3622110EA
COMPARE
0.26 217.5 115
₹207,000.00INR / Each
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 2 L/hr, 10 bar, Wall-Mt; 115V 5602241 Mfr # PLX3608010EA
COMPARE
0.53 145 115
₹246,000.00INR / Each
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 5 L/hr, 7 bar, Wall-Mt; 115V 5602242 Mfr # PLX3608610EA
COMPARE
1.32 101.5 115
₹207,000.00INR / Each
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 8 L/hr, 10 bar, Wall-Mt; 115V 5602244 Mfr # PLX3622710EA
COMPARE
2.11 145 115
₹200,000.00INR / Each
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 15 L/hr, 4 bar, Wall-Mt; 115V 5602245 Mfr # PLX3623010EA
COMPARE
3.96 58 115
₹234,000.00INR / Each
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 20 L/hr, 3 bar, Wall-Mt; 115V 5602246 Mfr # PLX3609510EA
COMPARE
5.28 43.5 115
₹246,000.00INR / Each
Etatron DLX-pH-RX/MBB DLX pH/ORP Pump Control System, 8 L/hr, 10 bar, Wall-Mt; 230V 5602284 Mfr # PLX3622810EA
COMPARE
2.11 145 230
₹224,000.00INR / Each
Showing 1- 7 of 7