Environmental Express® Zero Oxygen Solution
Accurate Dissolved oxygen results in any testing environment
  • Calibrate DO meters for measurement accuracy of dissolved oxygen
  • Manufactured according to the APHA and ASTM formulation using high-purity, oxygen-free water
Environmental Express zero oxygen standard solution ensures accuracy of your dissolved oxygen meter. For accurate dissolved oxygen blanking, the solution undergoes testing using the Winkler method. Follow all precautions recommended by your electrode manufacturer to ensure acceptable results.
Chemical oxygen scrubbers remove oxygen from the solution to provide fast calibration results.
4,800.00INR / Pint of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Volume (mL)
Availability Pricing
COMPARE
500
₹4,800.00INR / Pint of 1