Environmental Express® Washed and Dried TSS/VSS Glass Fiber Filters
Ready to be weighed and used for suspended solids filtration
  • Use in Standard Methods 2540
  • Provide consistently stable weights
  • Made from binderless borosilicate glass and have a 1.5 µm pore size
Laser cutting ensures a clean, sealed, fiber-free edge that maintains its integrity throughout washing and drying, as well as your testing process. These filters are rinsed three times with DI water to ensure they are fiber free. TSS filters are then dried in an oven at 105°C to obtain stable weights. VSS filters are dried at 550°C to ensure all volatiles are removed.
12,700.00 - ₹30,500.00INR / Pkg of 100
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Description
Availability Pricing
COMPARE
24 Washed and Dried Glass Fiber Filters, TSS, 24 mm; 100/Pk
₹12,700.00INR / Pkg of 100
COMPARE
34 Washed and Dried Glass Fiber Filters, TSS, 34 mm; 100/Pk
₹13,800.00INR / Pkg of 100
COMPARE
43 Washed and Dried Glass Fiber Filters, TSS, 43 mm; 100/Pk
₹14,300.00INR / Pkg of 100
COMPARE
47 Washed and Dried Glass Fiber Filters, TSS, 47 mm; 100/Pk
₹15,400.00INR / Pkg of 100
COMPARE
70 Washed and Dried Glass Fiber Filters, TSS, 70 mm; 100/Pk
₹15,600.00INR / Pkg of 100
COMPARE
90 Washed and Dried Glass Fiber Filters, TSS, 90 mm; 100/Pk
₹19,100.00INR / Pkg of 100
COMPARE
110 Washed and Dried Glass Fiber Filters, TSS, 110 mm; 100/Pk
₹24,700.00INR / Pkg of 100
COMPARE
125 Washed and Dried Glass Fiber Filters, TSS, 125 mm; 100/Pk
₹30,500.00INR / Pkg of 100
COMPARE
47 Washed and Dried Glass Fiber Filters, VSS, 47 mm; 100/Pk
₹17,200.00INR / Pkg of 100
COMPARE
90 Washed and Dried Glass Fiber Filters, VSS, 90 mm; 100/Pk
₹21,900.00INR / Pkg of 100
COMPARE
110 Washed and Dried Glass Fiber Filters, VSS, 110 mm; 100/Pk
₹27,600.00INR / Pkg of 100
Showing 1- 11 of 11