Environmental Express® TrustEE™ Sample Collection Bottles
TrustEE™ sample collection bottles for drinking water and wastewater sampling
  • Certified for environmental compliance testing
  • Interference-free composition
  • Applicable to water and wastewater testing workflows
TrustEE™ bottles are composed of (interference-free) high-density polyethylene with a polypropylene unlined cap conforming to EPA sample container requirements. They are screened at routine method MQL requirements to eliminate interference concerns from sampling point as a source rerun or resampling that can extend testing schedules.
These bottles are provided specifically for wastewater and drinking water testing methods and are level 1 certified to comply with EPA guidance for sample containers. They also comply with requirements for common methods used in PFAS (Per- and polyfluorinated alkyl substances) analysis including EPA methods 533, 537, and 1633.
Available in volumes of 125, 250, and 500 mL.
10,200.00 - ₹47,500.00INR / Pkg of 48
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Level Number Description
Availability Pricing
COMPARE
125 1 Sample Collection Bottle, Level 1, HDPE, 125 mL; 72/PK
₹10,200.00INR / Pkg of 72
COMPARE
250 1 Sample Collection Bottle, Level 1, HDPE, 250 mL; 72/PK
₹14,100.00INR / Pkg of 72
COMPARE
250 1 Sample Collection Bottle, Level 1, HDPE, 250 mL; 250/PK
₹47,500.00INR / Pkg of 250
COMPARE
500 1 Sample Collection Bottle, Level 1, HDPE, 500 mL; 48/PK
₹12,200.00INR / Pkg of 48
COMPARE
500 1 Sample Collection Bottle, Level 1, HDPE, 500 mL; 125/PK
₹31,000.00INR / Pkg of 125