Environmental Express® Series 1500 Probes
pH and conductivity probes for use with 1500 series meters
  • pH probe measurement range from 0 to 14
  • Conductivity probe measurement range from 0.1 mS/m to 10 S/m
In addition, pH and conductivity probes both offer temperature measurement.
23,400.00 - ₹34,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
All-in-One pH Electrode
₹23,400.00INR / Each
COMPARE
Conductivity Probe
₹34,900.00INR / Each