Environmental Express® Round Jugs, HDPE
Easily carry with the molded-in handle
  • Lightweight design won't add unnecessary weight
  • Leakproof cap keeps materials contained
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Level 3 containers meet EPA guidelines as well, but do not ship with a certificate of analysis—ideal for general-use
Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE.
3,000.00 - ₹42,700.00INR / Case of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Level Number
Availability Pricing
COMPARE
2 1
₹3,000.00INR / Case of 6
COMPARE
4 1
₹3,300.00INR / Case of 6
COMPARE
4 1
₹39,400.00INR / Case of 50
COMPARE
2 3
₹4,500.00INR / Case of 6
COMPARE
2 3
₹42,700.00INR / Case of 84
COMPARE
4 3
₹3,900.00INR / Case of 6