Environmental Express® QuikVap® Evaporating System
One system handles both flask and pan analysis methods
  • Accepts both borosilicate glass flasks or aluminum evaporating pans--simply switch out adapter plate
  • Evaporate n-hexane quickly and effectively for oil and grease testing
  • Caution: Operation should take place under an operating fume hood
Versatile system evaporates n-hexane quickly and effectively with adjustable temperature control, airflow control, and vessel configuration. Use digital controller to accurately set and maintain temperature throughout the evaporation process. System accepts either 100-mL flasks or aluminum pans with 70 or 105 mm diameter.
Connect to dry air or nitrogen source, and set temperature to 65°C. A sample of 40 mL of solvent will take approximately 20 minutes to evaporate.
Caution: Operation should take place under an operating fume hood.
508,000.00 - ₹755,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
₹508,000.00INR / Each
COMPARE
240
₹755,000.00INR / Each