Environmental Express® ProWeigh® and ProWeigh® Double Filters
Decrease sample prep time for your suspended solids testing
  • Compliant with Standard Methods 2540 Ready-to-use filters for total and volatile suspended solids Each filter is individually prepared, weighed, and labeled to be ready for use upon receipt Eliminate loose fibers and inaccurate weights with precise, laser-cut edges Quality control ensures filter integrity with visual inspection after each cutting, washing, and drying step
Filters for TSS or VSS.
20,100.00 - ₹45,500.00INR / Pkg of 100
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (micron) Description
Availability Pricing
COMPARE
76 1.5 Double Weigh Filters, VSS, 76 mm; 100Pk
₹31,000.00INR / Pkg of 100
COMPARE
32 1.5 Single Weigh Filters, TSS, 32 mm; 100/Pk
₹20,100.00INR / Pkg of 100
COMPARE
42 1.5 Filters, TSS, 42 mm; 100/Pk
₹21,700.00INR / Pkg of 100
COMPARE
47 1.5 Filters, TSS, 47 mm; 100/Pk
₹20,500.00INR / Pkg of 100
COMPARE
55 1.5 Single Weigh Filters, TSS, 55 mm; 100/Pk
₹22,200.00INR / Pkg of 100
COMPARE
70 1.5 Single Weigh Filters, TSS, 70 mm; 100/Pk
₹31,500.00INR / Pkg of 100
COMPARE
76 1.5 Filters, TSS, 76 mm; 100/Pk
₹32,000.00INR / Pkg of 100
COMPARE
90 1.5 Single Weigh Filters, TSS, 90 mm; 100/Pk
₹33,700.00INR / Pkg of 100
COMPARE
110 1.5 Single Weigh Filters, TSS, 110 mm; 100/Pk
₹42,900.00INR / Pkg of 100
COMPARE
42 1.5 Single Weigh Filters, VSS, 42 mm; 100/Pk
₹24,800.00INR / Pkg of 100
COMPARE
47 1.5 Filters, VSS, 47 mm; 100/Pk
₹23,000.00INR / Pkg of 100
COMPARE
55 1.5 Single Weigh Filters, VSS, 55 mm; 100/Pk
₹24,500.00INR / Pkg of 100
COMPARE
70 1.5 Single Weigh Filters, VSS, 70 mm; 100/Pk
₹33,700.00INR / Pkg of 100
COMPARE
76 1.5 Single Weigh Filters, VSS, 76 mm; 100/Pk
₹34,200.00INR / Pkg of 100
COMPARE
90 1.5 Filters, VSS, 90 mm; 100/Pk
₹36,000.00INR / Pkg of 100
COMPARE
110 1.5 Single Weigh Filters, VSS, 110 mm; 100/Pk
₹45,500.00INR / Pkg of 100
COMPARE
47 1.5 Double Weigh Filters, TSS, 47 mm; 100/Pk
₹31,700.00INR / Pkg of 100
COMPARE
76 1.5 Double Weigh Filters, TSS, 76 mm; 100/Pk
₹42,400.00INR / Pkg of 100
COMPARE
90 1.5 Double Weigh Filters, TSS, 90 mm; 100/Pk
₹43,200.00INR / Pkg of 100
COMPARE
47 1.5 Double Weigh Filters, VSS, 47 mm; 100/Pk
₹34,300.00INR / Pkg of 100
Showing 1- 20 of 20