Environmental Express® pH 2.00 Buffer Solution
Calibrate pH instruments to ensure accuracy
  • Lab-grade buffers available in multiple volumes to reduce waste and support various levels of throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST/USA buffer value of pH 2.00
pH 2.00 is a low-range pH meter calibration point. All Environmental Express® pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations.
If unopened, shelf life of pH 2.00 is 2 years; once opened, use within 90 days.
5,400.00 - ₹17,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL)
Availability Pricing
COMPARE
500
₹5,400.00INR / Each
COMPARE
1000
₹8,700.00INR / Each
COMPARE
3800
₹17,300.00INR / Each