Environmental Express® Oblong Bottles, HDPE
Store and retrieve samples with ease
  • Oblong shape minimizes dead space between bottles when stored
  • Keep samples safe with leakproof cap
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Level 3 containers meet EPA guidelines as well, but do not ship with a certificate
4,700.00 - ₹99,900.00INR / Case of 48
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Level Number Special Features
Availability Pricing
COMPARE
125 1
₹10,200.00INR / Case of 48
COMPARE
125 1
₹99,900.00INR / Case of 500
COMPARE
250 1
₹55,100.00INR / Case of 250
COMPARE
250 1
₹4,700.00INR / Case of 24
COMPARE
250 3
₹52,200.00INR / Case of 250
COMPARE
500 1
₹7,700.00INR / Case of 24
COMPARE
500 1
₹52,800.00INR / Case of 160
COMPARE
500 3
₹50,200.00INR / Case of 160
COMPARE
1000 1
₹58,900.00INR / Case of 116
COMPARE
1000 1 HCl Rinsed
₹54,300.00INR / Case of 116
COMPARE
1000 1 Nitric Rinsed
₹52,800.00INR / Case of 116
COMPARE
1000 1
₹7,400.00INR / Case of 12
COMPARE
1000 3
₹40,800.00INR / Case of 116
Showing 1- 13 of 13