Environmental Express® Oakton® 300 Series Multiparameter Smart Handheld Meters Accessories

Optimize performance of your Oakton Smart meters with accessories and replacement parts
  • Suitable for use with Oakton 300 series smart meters and meter kits
For your convenience, calibration standards, sensor heads, and sensor cartridges are available for your Oakton meters.
5,600.00 - ₹140,000.00INR / Each

11 variations of this product are available.

See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Sensor Head; 2-m Cable
₹44,900.00INR / Each
COMPARE
EC Sensor Head; 2-m Cable
₹36,400.00INR / Each
COMPARE
pH Sensor Cartridge, Double-Junction
₹33,700.00INR / Each
COMPARE
EC Sensor Cartridge, 4-Cell
₹43,700.00INR / Each
COMPARE
Optical DO Sensor; 2-m Cable
₹131,000.00INR / Each
COMPARE
Optical DO Sensor; 5-m Cable
₹140,000.00INR / Each
COMPARE
Replacement DO Sensor Cap
₹22,800.00INR / Each
COMPARE
pH Sensor Head; 5-m Cable
₹49,400.00INR / Each
COMPARE
EC Sensor Head; 5-m Cable
₹40,000.00INR / Each
COMPARE
Buffer Pack, pH 4.01, 7.00, and 10.01; 60-mL of each
₹6,000.00INR / Each
COMPARE
Conductivity and TDS Standards Pack, 84 µS, 1413 µS, 12.88 mS, and 111.8 mS; 60-mL of each
₹5,600.00INR / Pkg of 4
Showing 1- 11 of 11

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat