Environmental Express® AirLite™ Benchtop Enclosure
Maintain clean air for more uniform digestions
  • Get HEPA-filtered air just about anywhere!
  • Exhaust may be connected to existing ductwork
  • View digestions—clear PETG construction on all sides
  • Easily access chamber—door opens a full 180°
  • Cleanup is a breeze with removable, chemical-resistant bottom
  • Quiet, easily accessible 117 cfm PVC coated fan
  • Change HEPA filter in minutes
This enclosure is compatible with standard HotBlock® Digestion Systems.
656,000.00 - ₹670,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exhaust Volume (CFM) Power (VAC)
Availability Pricing
COMPARE
117 240
₹670,000.00INR / Each
COMPARE
120
₹656,000.00INR / Each