Elmi Sky Line Orbital Shakers
Optimal amplitude of platform rotation for intensive mixing
  • Variable speed control for gentle or high agitation
  • Continuous-duty operation
Elmi orbital shakers provide excellent rotational speed control at an economical price. Great selection of accessories make it easy to adapt the shakers to a variety of chemical, biological, and other laboratory applications.
A wide selection of accessories are available to accommodate different mixing vessels. Use the magnetic clips to hold your bottles and flasks in place. It even accommodates round-bottom flasks. Universal roller clip adapter is ideal for shaking vessels of different sizes at once.
200,000.00 - ₹296,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Elmi Sky Line Digital Orbital Shaker, Large, 10 mm , 115/230 VAC WW-51710-01 Mfr # SHKR-01
COMPARE
Digital Orbital Shaker, Large, 10 mm , 115/230 VAC
₹296,000.00INR / Each
Elmi Sky Line Digital Orbital Shaker, Large, 20 mm Amplitude, 115/230 VAC WW-51710-03 Mfr # SHKR-02
COMPARE
Digital Orbital Shaker, Large, 20 mm Amplitude, 115/230 VAC
₹296,000.00INR / Each
Elmi Sky Line Digital Orbital Shaker, 10 mm Amplitude, 115/230 VAC WW-51710-07 Mfr # SHKR-03
COMPARE
Digital Orbital Shaker, , 10 mm Amplitude, 115/230 VAC
₹278,000.00INR / Each
Elmi Sky Line Digital Orbital Shaker, Medium, 20 mm Amplitude, 115/230 VAC WW-51710-09 Mfr # SHKR-04
COMPARE
Digital Orbital Shaker, Medium, 20 mm Amplitude, 115/230 VAC
₹278,000.00INR / Each
Elmi Sky Line Digital Orbital Shaker, Small, 20 mm Amplitude, 115/230 VAC WW-51710-13 Mfr # SHKR-06
COMPARE
Digital Orbital Shaker, Small, 20 mm Amplitude, 115/230 VAC
₹200,000.00INR / Each
Elmi Sky Line Analog Orbital Shaker, Large, 10 mm Amplitude, 115/230 VAC WW-51710-17 Mfr # SHKR-07
COMPARE
Analog Orbital Shaker, Large, 10 mm Amplitude, 115/230 VAC
₹233,000.00INR / Each
Elmi Sky Line Analog Orbital Shaker, Large, 20 mm Amplitude, 115/230 VAC WW-51710-19 Mfr # SHKR-08 (s-3.02 20L)
COMPARE
Analog Orbital Shaker, Large, 20 mm Amplitude, 115/230 VAC
₹233,000.00INR / Each
Elmi Sky Line Analog Orbital Shaker, Medium, 10 mm Amplitude, 115/230 VAC WW-51710-21 Mfr # SHKR-09
COMPARE
Analog Orbital Shaker, Medium, 10 mm Amplitude, 115/230 VAC
₹219,000.00INR / Each
Elmi Sky Line ORBTL SHKR ANLG SHKR-10 115/233 WW-51710-23 Mfr # S.302 20M
COMPARE
Analog Orbital Shaker, Medium, 20 mm Amplitude, 115/230 VAC
₹219,000.00INR / Each
Showing 1- 9 of 9