Eldon James Threaded NPT(M) to Hose Barb Fittings, 90° Elbow, Natural Nylon
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. The elbows feature a 90° bend and is designed to fit in a tight space. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
2,000.00 - ₹2,900.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Flow Pattern Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" NPT(M) x 5/8" ID; 10/Pk 5012584 Mfr # L12-10NN
COMPARE
5/8 90° elbow 3/4
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" NPT(M) x 3/4" ID; 10/Pk 5012585 Mfr # L12-12NN
COMPARE
3/4 90° elbow 3/4
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" NPT(M) x 1" ID; 10/Pk 5012586 Mfr # L12-16NN
COMPARE
1 90° elbow 3/4
₹2,900.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" NPT(M) x 1/4" ID; 10/Pk 5012587 Mfr # L12-4NN
COMPARE
1/4 90° elbow 3/4
₹2,000.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" NPT(M) x 3/8" ID; 10/Pk 5012588 Mfr # L12-6NN
COMPARE
3/8 90° elbow 3/4
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" NPT(M) x 1/2" ID; 10/Pk 5012589 Mfr # L12-8NN
COMPARE
1/2 90° elbow 3/4
₹2,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 1" NPT(M) x 1" ID; 10/Pk 5012590 Mfr # L16-16NN
COMPARE
1 90° elbow 1
₹2,900.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Nylon, 1/2" NPT(M) x 3/4" ID; 10/Pk 5012684 Mfr # L16-12NN
COMPARE
3/4 90° elbow 1/2
₹2,900.00INR / Pkg of 10
Showing 1- 8 of 8