Eldon James SeriesLock® Quick-Disconnect Fittings, Hose Barb Body & Insert, X-Large Housing (1" Barb), Polysulfone
Specially designed quick-disconnect fitting provides a high-volume flow at low line pressures
  • Spring-free flow path® design
  • Audible click ensures secure connection
  • Safety slide release minimizes accidental disconnects
  • Sterile quick disconnect
  • High flow yet compact to fit in tight spaces
  • Superior barb design provides a leak-free connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
The SeriesLock® quick disconnect fittings with spring-free fluid path allow a high-volume flow while minimizing turbulence, foaming, and bubbles. For a secure, leak-free connection, push an insert into a fitting body until you hear an audible “click.” Simply use the safety slide release to disconnect the fittings. The slide release eliminates accidental separation while offering easy disconnect with one hand even while wearing gloves. Superior barb design prevents any parting lines on the barb surface, eliminating any potential leak point. It also allows the tubing to relax past the barb and a space for a clamp. Recessed connection features protect contact surfaces and prolong the life of the O-rings.
Constructed of translucent polysulfone, fittings show excellent resistance to mineral acids, akali and salt solutions, detergents, and hydrocarbon oils. These durable fittings are rated for continuous use at temperatures up to 149°C (300°F).
Note: Use X-large polysulfone inserts only with X-large polysulfone bodies.
9,900.00 - ₹22,500.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Connection Type
Availability Pricing
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, Silicone O-Ring, SS Spring, X-Large, 1" ID 5013837 Mfr # B-FV-SO-SS-16
COMPARE
Quick-Disconnect Body x Hose Barb
₹17,100.00INR / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, Silicone O-Ring, SS Spring, X-Large, 1" ID 5013846 Mfr # B-MV-S-SS-16
COMPARE
Quick-Disconnect Insert x Hose Barb
₹16,800.00INR / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Non-Valved, Silicone O-Ring, SS Spring, X-Large, 1" ID 5013852 Mfr # B-FO-SO-SS-16
COMPARE
Quick-Disconnect Body x Hose Barb
₹13,000.00INR / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, X-Large, 1" ID 5013858 Mfr # B-MO-S-16
COMPARE
Quick-Disconnect Insert x Hose Barb
₹9,900.00INR / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Insert, Panel Mount, Polysulfone, Valved, Silicone O-Ring, SS Spring, X-Large, 1" ID 5013890 Mfr # PM-MV-S-SS-16
COMPARE
Quick-Disconnect Panel-Mount Insert x Hose Barb
₹22,500.00INR / Each