Eldon James Hose Barb to Plug Fittings, Straight Adapter, Animal-Derivative-Free Polypropylene
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Simply insert the hose barb plug in the soft tubing to stop the fluid flow. Each fitting features a single barb on one end, ensuring a tight seal to keep the tubing securely in place.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. However, it’s important to avoid using them with oxidizing agents, as these can compromise their integrity.
1,000.00 - ₹3,300.00INR / Pkg of 10
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/PK 5012729 Mfr # P0-1.5-200PP
COMPARE
3/32 Polypropylene Hose Barb x Plug
₹1,000.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 5/8" ID; 10/PK 5012730 Mfr # P0-10PP
COMPARE
5/8 Polypropylene Hose Barb x Plug
₹3,300.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/16" ID; 10/PK 5012731 Mfr # P0-1-200PP
COMPARE
1/16 Polypropylene Hose Barb x Plug
₹1,000.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 5/32" ID; 10/PK 5012732 Mfr # P0-2.5-200PP
COMPARE
5/32 Polypropylene Hose Barb x Plug
₹1,300.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/8" ID; 10/PK 5012733 Mfr # P0-2-200PP
COMPARE
1/8 Polypropylene Hose Barb x Plug
₹1,200.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/4" ID; 10/PK 5012734 Mfr # P0-4PP
COMPARE
1/4 Polypropylene Hose Barb x Plug
₹2,200.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/8" ID; 10/PK 5012735 Mfr # P0-6PP
COMPARE
3/8 Polypropylene Hose Barb x Plug
₹2,300.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 1/2" ID; 10/PK 5012736 Mfr # P0-8PP
COMPARE
1/2 Polypropylene Hose Barb x Plug
₹2,300.00INR / Pkg of 10
Eldon James Hose Barb to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, 3/16" ID; 10/PK 5012867 Mfr # P0-3-200PP
COMPARE
3/16 Polypropylene Hose Barb x Plug
₹1,600.00INR / Pkg of 10
Showing 1- 9 of 9