Eldon James Hose Barb Fittings, Y Reducer, Natural Nylon
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. They allow for easy fluid flow combination or splitting with reduction Y fittings, which have one large hose barb and two smaller ones. Each fitting features a single barb on each end, ensuring a tight seal to keep the tubing securely in place.
An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These natural nylon fittings offer excellent tensile strength, stiffness, and impact resistance, making them suitable for various applications. Their resistance to fuels and lubricants is a definite advantage in many industrial settings. However, it’s important to avoid using them with phenols, strong acids, and oxidizing agents, as these can compromise their integrity.
3,400.00 - ₹6,500.00INR / Pkg of 10
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 5/8" ID x 3/4" ID; 10/PK 5012602 Mfr # RY10-12NN
COMPARE
3/4 5/8 5/8
₹5,900.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 3/4" ID x 1" ID; 10/PK 5012603 Mfr # RY12-16NN
COMPARE
1 3/4 3/4
₹6,500.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 1/8" ID x 1/4" ID; 10/PK 5012604 Mfr # RY2-4NN
COMPARE
1/4 1/8 1/8
₹3,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 3/16" ID x 5/16" ID; 10/PK 5012605 Mfr # RY3-5NN
COMPARE
5/16 3/16 3/16
₹4,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 1/4" ID x 3/8" ID; 10/PK 5012606 Mfr # RY4-6NN
COMPARE
3/8 1/4 1/4
₹4,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 3/8" ID x 1/2" ID; 10/PK 5012607 Mfr # RY6-8NN
COMPARE
1/2 3/8 3/8
₹3,900.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Nylon, 1/2" ID x 5/8" ID; 10/PK 5012608 Mfr # RY8-10NN
COMPARE
5/8 1/2 1/2
₹5,600.00INR / Pkg of 10
Showing 1- 7 of 7