Eldon James Hose Barb Fittings, Straight Reducer, Natural Nylon
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily connect two different size tubing with straight reducers. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These natural nylon fittings offer excellent tensile strength, stiffness, and impact resistance, making them suitable for various applications. Their resistance to fuels and lubricants is a definite advantage in many industrial settings. However, it’s important to avoid using them with phenols, strong acids, and oxidizing agents, as these can compromise their integrity.
1,000.00 - ₹2,500.00INR / Pkg of 10
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 5/8" ID x 1/4" ID; 10/PK 5012541 Mfr # C10-4NN
COMPARE
5/8 1/4 Nylon
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 5/8" ID x 5/16" ID; 10/PK 5012542 Mfr # C10-5NN
COMPARE
5/8 5/16 Nylon
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 5/8" ID x 3/8" ID; 10/PK 5012543 Mfr # C10-6NN
COMPARE
5/8 3/8 Nylon
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 5/8" ID x 1/2" ID; 10/PK 5012544 Mfr # C10-8NN
COMPARE
5/8 1/2 Nylon
₹2,200.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 3/4" ID x 5/8" ID; 10/PK 5012545 Mfr # C12-10NN
COMPARE
5/64 5/8 Nylon
₹2,500.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 3/4" ID x 3/8" ID; 10/PK 5012546 Mfr # C12-6NN
COMPARE
5/64 3/8 Nylon
₹2,200.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 3/4" ID x 1/2" ID; 10/PK 5012547 Mfr # C12-8NN
COMPARE
5/64 1/2 Nylon
₹2,200.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1" ID x 5/8" ID; 10/PK 5012548 Mfr # C16-10NN
COMPARE
1 5/8 Nylon
₹2,500.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1" ID x 3/4" ID; 10/PK 5012549 Mfr # C16-12NN
COMPARE
1 3/4 Nylon
₹2,500.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1" ID x 1/2" ID; 10/PK 5012550 Mfr # C16-8NN
COMPARE
1 1/2 Nylon
₹2,500.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 5/32" ID x 3/32" ID; 10/PK 5012551 Mfr # C2.5-1.5-200NN
COMPARE
5/32 3/32 Nylon
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 5/32" ID x 1/8" ID; 10/PK 5012552 Mfr # C2.5-2NN
COMPARE
5/32 1/8 Nylon
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1/8" ID x 3/32" ID; 10/PK 5012553 Mfr # C2-1.5-200NN
COMPARE
1/8 3/32 Nylon
₹1,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1/8" ID x 1/16" ID; 10/PK 5012554 Mfr # C2-1-200NN
COMPARE
1/8 1/16 Nylon
₹1,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 3/16" ID x 3/32" ID; 10/PK 5012555 Mfr # C3-1.5-200NN
COMPARE
3/16 3/32 Nylon
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 3/16" ID x 1/16" ID; 10/PK 5012556 Mfr # C3-1-200NN
COMPARE
3/16 1/16 Nylon
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 3/16" ID x 1/8" ID; 10/PK 5012557 Mfr # C3-2NN
COMPARE
3/16 1/8 Nylon
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1/4" ID x 1/16" ID; 10/PK 5012558 Mfr # C4-1-200NN
COMPARE
1/4 1/16 Nylon
₹2,300.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1/4" ID x 5/32" ID; 10/PK 5012559 Mfr # C4-2.5NN
COMPARE
1/4 5/32 Nylon
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Nylon, 1/4" ID x 1/8" ID; 10/PK 5012560 Mfr # C4-2NN
COMPARE
1/4 1/8 Nylon
₹1,800.00INR / Pkg of 10
Showing 1 - 20 of 31