Eldon James Hose Barb Fittings, Straight Reducer, Natural Kynar®
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Biopharma compatible and USP 661 and BPOG compliant
Use these high-quality PVC-free fittings for biomedical research, pharmaceuticals, electronics, and general-purpose applications. Easily connect different-sized tubing with straight reducers. Each fitting features a single barb on either end, ensuring a tight seal to keep the tubing securely in place.
These natural Kynar® fittings offer excellent resistance to chemicals, flames, gamma radiation, UV light, and weather. They also demonstrate strong mechanical properties, low moisture absorption, and good machinability.
1,600.00 - ₹8,900.00INR / Pkg of 10
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 5/8" ID x 1/4" ID; 10/PK 5012509 Mfr # C10-4NK7
COMPARE
5/8 1/4 PVDF
₹5,300.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 5/8" ID x 5/16" ID; 10/PK 5012510 Mfr # C10-5NK7
COMPARE
5/8 5/16 PVDF
₹5,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 5/8" ID x 3/8" ID; 10/PK 5012511 Mfr # C10-6NK7
COMPARE
5/8 3/8 PVDF
₹5,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 5/8" ID x 1/2" ID; 10/PK 5012512 Mfr # C10-8NK7
COMPARE
5/8 1/2 PVDF
₹5,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/4" ID x 5/8" ID; 10/PK 5012513 Mfr # C12-10NK7
COMPARE
3/4 5/8 PVDF
₹7,000.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/4" ID x 3/8" ID; 10/PK 5012514 Mfr # C12-6NK7
COMPARE
3/4 3/8 PVDF
₹6,300.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/4" ID x 1/2" ID; 10/PK 5012515 Mfr # C12-8NK7
COMPARE
3/4 1/2 PVDF
₹6,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1" ID x 5/8" ID; 10/PK 5012516 Mfr # C16-10NK7
COMPARE
1 5/8 PVDF
₹8,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1" ID x 3/4" ID; 10/PK 5012517 Mfr # C16-12NK7
COMPARE
1 3/4 PVDF
₹8,900.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1" ID x 1/2" ID; 10/PK 5012518 Mfr # C16-8NK7
COMPARE
1 1/2 PVDF
₹8,100.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 5/32" ID x 3/32" ID; 10/PK 5012519 Mfr # C2.5-1.5-200NK7
COMPARE
5/32 3/32 PVDF
₹2,400.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 5/32" ID x 1/8" ID; 10/PK 5012520 Mfr # C2.5-2NK7
COMPARE
5/32 1/8 PVDF
₹2,900.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1/8" ID x 3/32" ID; 10/PK 5012521 Mfr # C2-1.5-200NK7
COMPARE
1/8 3/32 PVDF
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1/8" ID x 1/16" ID; 10/PK 5012522 Mfr # C2-1-200NK7
COMPARE
1/8 1/16 PVDF
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/16" ID x 3/32" ID; 10/PK 5012523 Mfr # C3-1.5-200NK7
COMPARE
3/16 3/32 PVDF
₹2,700.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/16" ID x 1/16" ID; 10/PK 5012524 Mfr # C3-1-200NK7
COMPARE
3/16 1/16 PVDF
₹2,300.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/16" ID x 5/32" ID; 10/PK 5012525 Mfr # C3-2.5-200NK7
COMPARE
3/16 5/32 PVDF
₹2,700.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 3/16" ID x 1/8" ID; 10/PK 5012526 Mfr # C3-2NK7
COMPARE
3/16 1/8 PVDF
₹2,900.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1/4" ID x 3/32" ID; 10/PK 5012527 Mfr # C4-1.5-200NK7
COMPARE
1/4 3/32 PVDF
₹3,700.00INR / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Natural Kynar®, 1/4" ID x 1/16" ID; 10/PK 5012528 Mfr # C4-1-200NK7
COMPARE
1/4 1/16 PVDF
₹3,400.00INR / Pkg of 10
Showing 1 - 20 of 33