Eldon James Hose Barb Fittings, 90° Elbow, Stainless Steel
Easily change the direction of the fluid flow with 90° elbows
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. The elbows feature a 90° bend and is designed to fit in a tight space. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
2,500.00 - ₹11,600.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 5/8" ID; 1/Each 5012779 Mfr # L0-10SS
COMPARE
5/8 5/8 Hose Barb Elbow
₹9,500.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 3/4" ID; 1/Each 5012780 Mfr # L0-12SS
COMPARE
3/4 3/4 Hose Barb Elbow
₹11,600.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 5/32" ID; 1/Each 5012781 Mfr # L0-2.5SS
COMPARE
5/32 5/32 Hose Barb Elbow
₹2,500.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 1/8" ID; 1/Each 5012782 Mfr # L0-2SS
COMPARE
1/8 1/8 Hose Barb Elbow
₹3,100.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 3/16" ID; 1/Each 5012783 Mfr # L0-3SS
COMPARE
3/16 3/16 Hose Barb Elbow
₹3,100.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 1/4" ID; 1/Each 5012784 Mfr # L0-4SS
COMPARE
1/4 1/4 Hose Barb Elbow
₹4,500.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 5/16" ID; 1/Each 5012785 Mfr # L0-5SS
COMPARE
5/16 5/16 Hose Barb Elbow
₹5,200.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 3/8" ID; 1/Each 5012786 Mfr # L0-6SS
COMPARE
3/8 3/8 Hose Barb Elbow
₹6,600.00INR / Each
Eldon James Hose Barb Fitting, 90° Elbow, Stainless Steel, 1/2" ID; 1/Each 5012787 Mfr # L0-8SS
COMPARE
1/2 1/2 Hose Barb Elbow
₹7,600.00INR / Each
Showing 1- 9 of 9