Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon
Easily change the direction of the fluid flow with 90° elbows
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. The elbows feature a 90° bend and is designed to fit in a tight space. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
1,500.00 - ₹2,800.00INR / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/32" ID; 10/Pk 5012572 Mfr # L0-1.5-200NN
COMPARE
3/32 3/32 Hose Barb Elbow
₹1,500.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 5/8" ID; 10/Pk 5012573 Mfr # L0-10NN
COMPARE
5/8 5/8 Hose Barb Elbow
₹2,000.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 1/16" ID; 10/Pk 5012574 Mfr # L0-1-200NN
COMPARE
1/16 1/16 Hose Barb Elbow
₹1,500.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/4" ID; 10/Pk 5012575 Mfr # L0-12NN
COMPARE
3/4 3/4 Hose Barb Elbow
₹2,400.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 1" ID; 10/Pk 5012576 Mfr # L0-16NN
COMPARE
1 1 Hose Barb Elbow
₹2,800.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 5/32" ID; 10/Pk 5012577 Mfr # L0-2.5NN
COMPARE
5/32 5/32 Hose Barb Elbow
₹2,300.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 1/8" ID; 10/Pk 5012578 Mfr # L0-2-200NN
COMPARE
1/8 1/8 Hose Barb Elbow
₹1,600.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/16" ID; 10/Pk 5012579 Mfr # L0-3NN
COMPARE
3/16 3/16 Hose Barb Elbow
₹2,700.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 1/4" ID; 10/Pk 5012580 Mfr # L0-4NN
COMPARE
1/4 1/4 Hose Barb Elbow
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 5/16" ID; 10/Pk 5012581 Mfr # L0-5NN
COMPARE
5/16 5/16 Hose Barb Elbow
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 3/8" ID; 10/Pk 5012582 Mfr # L0-6NN
COMPARE
3/8 3/8 Hose Barb Elbow
₹1,800.00INR / Pkg of 10
Eldon James Hose Barb Fittings, 90° Elbow, Natural Nylon, 1/2" ID; 10/Pk 5012583 Mfr # L0-8NN
COMPARE
1/2 1/2 Hose Barb Elbow
₹2,200.00INR / Pkg of 10
Showing 1- 12 of 12