Eldex Optos Model 3 High-Pressure Piston Metering Pumps with Damper and Transducer
Compact pumps provide precise flow rate control and smooth fluid delivery at high pressures
  • Reciprocating piston design for precise flow control
  • High-resolution stepper motor minimizes pulsation
  • Damper/transducer provides smooth fluid delivery and pressure monitoring
  • Control flow remotely via analog or RS-232 connection
  • Integrated piston wash extends piston seal life
  • Solvent-resistant membrane keypad
  • Use with liquids with temperatures ranging from 5 to 260°C (41 to 500°F)
  • Compact meter only measures 10 cm W x 24 cm D (4” x 9.5”), saving valuable benchtop space
The Optos series metering pumps deliver high precision and reliability at high pressures. Achieve flow rates from 0.01 to 80 mL/min with ±0.3% reproducibility with Model 3 meters. The reciprocating piston offers precise flow control while an advanced software algorithms control high-resolution stepper motor to maximize flow output and minimize pulsation. Easily set flow rates using the up and down buttons on the solvent-resistant keypad. Remotely control your meter via an analog signal (4 to 20 mA or 0 to 5 VDC) or RS-232. Contact closures allow remote start and stop, flow output (0 to 5 VDC), and pump error output.
All pumps come with a pulse damper/transducer installed, connecting the pump outlet and the damper inlet. The integrated pulse damper/transducer further reduces pulsation for smooth flow as well as the ability to monitor pressure and set high- and low-pressure limits.
Select a pump that uses the upper half of the flow range listed for each pump. Piston cylinder comes available in two materials—316 stainless steel (SS) and PEEK. 316 SS offers excellent chemical and corrosion resistance; use PEEK for extremely corrosive fluids. A fully integrated piston wash system cleans out buffers or solutions which can crystallize and lead to abrasion of the piston seal, shortening the piston seal life.
573,000.00 - ₹644,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI)
Availability Pricing
Optos Model 3LMP Piston Metering Pump with Damper and Transducer, 316 SS, 3/32" Piston Dia, 0.01 to 10 mL/min 4853185 Mfr # 6354
COMPARE
10 3000
₹587,000.00INR / Each
Eldex Optos Model 3SMP Piston Metering Pump with Damper and Transducer, 316 SS, 1/8" Piston Dia, 0.01 to 20 mL/min 4853186 Mfr # 6355
COMPARE
20 1500
₹573,000.00INR / Each
Eldex Optos Model 3HMP Piston Metering Pump with Damper and Transducer, 316 SS, 1/4" Piston Dia, 0.04 to 80 mL/min 4853187 Mfr # 6137
COMPARE
80 750
₹644,000.00INR / Each