Eldex Optos Model 1 High-Pressure Piston Metering Pumps with Damper and Transducer
Compact pumps provide precise flow rate control and smooth fluid delivery at high pressures
  • Reciprocating piston design for precise flow control
  • High-resolution stepper motor minimizes pulsation
  • Damper/transducer provides smooth fluid delivery and pressure monitoring
  • Control flow remotely via analog or RS-232 connection
  • Integrated piston wash extends piston seal life
  • Solvent-resistant membrane keypad
  • Use with liquids with temperatures ranging from 5 to 260°C (41 to 500°F)
  • Compact meter only measures 10 cm W x 24 cm D (4” x 9.5”), saving valuable benchtop space
The Optos series metering pumps deliver high precision and reliability at high pressures. Achieve flow rates from 0.002 to 20 mL/min with ±0.3% reproducibility with Model 1 meters. The reciprocating piston offers precise flow control while an advanced software algorithms control high-resolution stepper motor to maximize flow output and minimize pulsation. Easily set flow rates using the up and down buttons on the solvent-resistant keypad. Remotely control your meter via an analog signal (4 to 20 mA or 0 to 5 VDC) or RS-232. Contact closures allow remote start and stop, flow output (0 to 5 VDC), and pump error output.
All pumps come with a pulse damper/transducer installed, connecting the pump outlet and the damper inlet. The integrated pulse damper/transducer further reduces pulsation for smooth flow as well as the ability to monitor pressure and set high- and low-pressure limits.
Select a pump that uses the upper half of the flow range listed for each pump. Piston cylinder comes available in two materials—316 stainless steel (SS) and PEEK. 316 SS offers excellent chemical and corrosion resistance; use PEEK for extremely corrosive fluids. A fully integrated piston wash system cleans out buffers or solutions which can crystallize and lead to abrasion of the piston seal, shortening the piston seal life.
573,000.00 - ₹683,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
Eldex Optos Model 1LMP Piston Metering Pump with Damper and Transducer, 316 SS, 3/32" Piston Dia, 0.002 to 2.5 mL/min 4853163 Mfr # 5985
COMPARE
2.5 6000 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
₹587,000.00INR / Each
Eldex Optos Model 1SMP Piston Metering Pump with Damper and Transducer, 316 SS, 1/8" Piston Dia, 0.003 to 5 mL/min 4853164 Mfr # 5967
COMPARE
5 6000 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE, Kal-Rez
₹573,000.00INR / Each
Optos Model 1HMP Piston Metering Pump with Damper and Transducer, 316 SS, 1/4" Piston Dia, 0.01 to 20 mL/min 4853165 Mfr # 6135
COMPARE
20 3000 316 stainless steel, sapphire, ruby, graphite fiber reinforced PTFE, PCTFE
₹644,000.00INR / Each
Eldex Optos Model 1LIP Piston Metering Pump with Damper and Transducer, PEEK, 3/32" Piston Dia, 0.002 to 2.5 mL/min 4853169 Mfr # 5965
COMPARE
2.5 4000 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
₹683,000.00INR / Each
Optos Model 1SIP Piston Metering Pump with Damper and Transducer, PEEK, 1/8" Piston Dia, 0.003 to 5 mL/min 4853170 Mfr # 5969
COMPARE
5 4000 PEEK, sapphire, ruby, UHMW polyethylene, PCTFE
₹683,000.00INR / Each