Edwards™ RV Series Rotary Vane Vacuum Pumps
Versatile rotary vane pumps are ideal for a wide array of applications
  • Pumps are two stage, oil-sealed, and sliding vane
  • Feature a gas-ballast control and mode selector that allows for six different control combinations
  • Two motor variants cover all voltage and frequency conditions
  • Choose pumps that are prepared to accept PFPE pump oil when working with high concentrations of oxygen
These pumps gives users options. The unique mode selector means that you only need one model for both high-throughput and high-vacuum applications. In addition to the mode selector, these pumps have a two-position gas ballast. The pumps are fast-acting and able to work with large amounts of vapor. These pumps are perfect for industrial use, chemical applications, sample preparation, mass spectrometry, electron microscopy, research & development, and high energy physics.
947,000.00 - ₹1,761,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Vacuum (torr) Power (VAC) Description
Availability Pricing
Edwards RV3 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 230 V for Europe) 7930074 Mfr # A65201903
COMPARE
1.5 x 10-3 Torr 115 / 230 RV3 rotary vane pump, speed at 50 Hz: 3.3 m3/hr, speed at 60 Hz: 3.9 m3/hr
₹947,000.00INR / Each
Edwards RV3 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930079 Mfr # A65201906
COMPARE
1.5 x 10-3 Torr 115 / 230 RV3 rotary vane pump, speed at 50 Hz: 3.3 m3/hr, speed at 60 Hz: 3.9 m3/hr
₹947,000.00INR / Each
Edwards RV3 Two Stage Rotary Vane Pump, PFPE-prepared; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930081 Mfr # A65209906
COMPARE
1.5 x 10-3 Torr, With PFPE oil: 1.5 x 10-2 Torr 115 / 230 RV3 rotary vane pump, speed at 50 Hz: 3.3 m3/hr, speed at 60 Hz: 3.9 m3/hr
₹1,269,000.00INR / Each
Edwards RV5 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 230 V for Europe) 7930082 Mfr # A65301903
COMPARE
1.5 x 10-3 Torr 115 / 230 RV5 rotary vane pump, speed at 50 Hz: 5.1 m3/hr, speed at 60 Hz: 6.2 m3/hr
₹1,134,000.00INR / Each
Edwards RV5 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930086 Mfr # A65301906
COMPARE
1.5 x 10-3 Torr 115 / 230 RV5 rotary vane pump, speed at 50 Hz: 5.1 m3/hr, speed at 60 Hz: 6.2 m3/hr
₹1,134,000.00INR / Each
Edwards RV5 Two Stage Rotary Vane Pump, PFPE-prepared; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930087 Mfr # A65309906
COMPARE
1.5 x 10-3 Torr, With PFPE oil: 1.5 x 10-2 Torr 115 / 230 RV5 rotary vane pump, speed at 50 Hz: 5.1 m3/hr, speed at 60 Hz: 6.2 m3/hr
₹1,455,000.00INR / Each
Edwards RV8 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 230 V for Europe) 7930088 Mfr # A65401903
COMPARE
1.5 x 10-3 Torr 115 / 230 RV8 rotary vane pump, speed at 50 Hz: 8.5 m3/hr, speed at 60 Hz: 10 m3/hr
₹1,304,000.00INR / Each
Edwards RV8 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930093 Mfr # A65401906
COMPARE
1.5 x 10-3 Torr 115 / 230 RV8 rotary vane pump, speed at 50 Hz: 8.5 m3/hr, speed at 60 Hz: 10 m3/hr
₹1,304,000.00INR / Each
Edwards RV8 Two Stage Rotary Vane Pump, PFPE-prepared; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930094 Mfr # A65409906
COMPARE
1.5 x 10-3 Torr, With PFPE oil: 1.5 x 10-2 Torr 115 / 230 RV8 rotary vane pump, speed at 50 Hz: 8.5 m3/hr, speed at 60 Hz: 10 m3/hr
₹1,625,000.00INR / Each
Edwards RV12 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 230 V for Europe) 7930095 Mfr # A65501903
COMPARE
1.5 x 10-3 Torr 115 / 230 RV12 rotary vane pump, speed at 50 Hz: 12 m3/hr, speed at 60 Hz: 14.2 m3/hr
₹1,440,000.00INR / Each
Edwards RV12 Two Stage Rotary Vane Pump; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930102 Mfr # A65501906
COMPARE
1.5 x 10-3 Torr 115 / 230 RV12 rotary vane pump, speed at 50 Hz: 12 m3/hr, speed at 60 Hz: 14.2 m3/hr
₹1,440,000.00INR / Each
Edwards RV12 Two Stage Rotary Vane Pump, PFPE-prepared; 115 to 230 V, 50/60 Hz, 1 phase (Factory set to 115 V for US) 7930103 Mfr # A65509906
COMPARE
1.5 x 10-3 Torr, With PFPE oil: 1.5 x 10-2 Torr 115 / 230 RV12 rotary vane pump, speed at 50 Hz: 12 m3/hr, speed at 60 Hz: 14.2 m3/hr
₹1,761,000.00INR / Each
Showing 1- 12 of 12