Dymo LabelWriter 450 Series Printers

Print labels from your PC or MAC
  • Direct thermal print technology eliminates ribbons, toner, and ink cartridges
  • Automatically checks and corrects U.S. addresses
Print crystal-clear text, graphics, and barcodes with 600 x 300 dpi resolution graphics. Printers allow you to choose from 60+ label styles and layouts just by browsing all available layouts on screen – quickly find the best label for any job. Label printers eliminate the hassle of sheet-fed labels by printing one label at a time, a few labels, or many labels in as little as two seconds each. Label printers handle many different label styles to accommodate numerous job functions — shipping and return labels, file folder labels, name badges, inventory, and bar code labels — and come with their own easy-to-use software that integrates seamlessly with Microsoft® Word or Outlook®, QuickBooks®, CardScan®, and other programs, without retyping. LabelWriter allows you to verify U.S. addresses and ZIP+4® codes with included Address Fixer™ software (requires Internet connection).

LabelWriter 450 prints basic labels for envelopes, packages, files inventory, bar codes, and more at speeds up to 51 labels per minute. Accepts LW labels.

LabelWriter 450 Twin Turbo holds two label rolls at once for maximum efficiency – print 60+ label layouts at speeds up to 71 labels per minute, without switching label rolls. Accepts LW labels.

LabelWriter 450 Duo prints 60+ paper label styles at speeds up to 71 labels per minute, and uses LW labels and D1 cassette labels.

System requirements: Windows® XP/Vista (32-bit, 64-bit) or Mac® OS 10.4 or higher
5,500.00 - ₹9,600.00INR / Pkg of 2

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Dymo 30327 File labels, 9/16" x 3 7/16", 130 labels/roll, 2 rolls/pack 6553010 Mfr # 30327
COMPARE
File labels, 1/2" x 3 1/2", 130 labels/roll, 2 rolls/pack
₹5,500.00INR / Pkg of 2
Dymo 30330 Return Address Labels, White, 3/4” X 2”-500 Per Roll,1 Roll Per Box 6553084 Mfr # 30330
COMPARE
Return Address Labels, White, 3/4” X 2”-500 Per Roll,1 Roll Per Box
₹5,600.00INR / Pkg of 1
Dymo 30336 Multi-purpose Labels, White, 1” X 2 1/8”-500 Per Roll,1 Roll Per Box 6553086 Mfr # 30336
COMPARE
Multi-purpose Labels, White, 1” X 2 1/8”-500 Per Roll,1 Roll Per Box
₹9,600.00INR / Pkg of 1
Dymo 30251 Address Label, 130 Labels Per Roll, Two Roll/Pack 6553385 Mfr # 30251
COMPARE
Address Label, 130 Labels Per Roll, Two Roll/Pack
₹6,300.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat