DWK Wheaton Culture Tubes with Screw Cap
Ideal for use with slant, shake, and drum roller cultures
  • Screw on cap provides secure seal
  • Autoclavable
These reusable culture tubes are manufactured from borosilicate glass 5.1 and feature a round-bottom design. Each tube comes with a 14B rubber lined screw on cap providing you with a secure seal.
59,100.00 - ₹143,000.00INR / Case of 144
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Length (mm) Tube Outside Diameter (OD) (mm)
Availability Pricing
DWK Life Sciences (Wheaton) Culture Tubes with White 14B Rubber Lined  Screw Cap, 16 x 100 mm; 144/CS 3459500 Mfr # 358606
COMPARE
11 100 16
₹59,100.00INR / Case of 144
DWK Life Sciences (Wheaton) Culture Tubes with White 14B Rubber Lined  Screw Cap, 16x 125 mm; 144/CS 3459501 Mfr # 358607
COMPARE
14 125 16
₹59,400.00INR / Case of 144
DWK Life Sciences (Wheaton) Culture Tubes with White 14B Rubber Lined Screw Cap, 20 x 150 mm; 144/CS 3459502 Mfr # 358610
COMPARE
27 150 20
₹82,600.00INR / Case of 144
DWK Life Sciences (Wheaton) Culture Tubes with PTFE Faced 14B Rubber Lined Screw Cap, 16 x 100 mm; 144/CS 3459503 Mfr # 358646
COMPARE
11 100 16
₹69,800.00INR / Case of 144
DWK Life Sciences (Wheaton) Culture Tubes with PTFE Faced 14B Rubber Lined Screw Cap, 16 x 125 mm; 144/CS 3459504 Mfr # 358647
COMPARE
14 125 16
₹71,100.00INR / Case of 144
DWK Life Sciences (Wheaton) Culture Tubes with PTFE Faced 14B Rubber Lined Screw Cap, 20 x 150 mm; 144/CS 3459505 Mfr # 358650
COMPARE
27 150 20
₹143,000.00INR / Case of 144