DWK Life Sciences (Kimble) Score-Break Ampules
Pre-scored stem permits safe and easy opening
  • Fabricated from USP Type I borosilicate glass
  • Excellent dimensional stability for high-speed filling
  • Stems can be pull or tip-sealed
  • Easy hermetic sealing
Manufactured from USP Type I borosilicate glass, these ampules conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1. They provide maximum solution stability and allow for easy hermetic sealing. Pre-scored constricted portion of the stem allows for safe and easy opening, eliminating the need for a special ampule file.
Methods of sealing include tip or pull. Short-stem ampules are tip-sealed, while trimmed stem ampules are pull-sealed.
17,200.00 - ₹227,000.00INR / Case of 144
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Color Tube Length (mm)
Availability Pricing
DWK Life Sciences (Kimble) Score-Break Ampule, Amber, Trimmed Stem, Gross Pack, 1 mL; 144/CS 3459506 Mfr # 12040G-1
COMPARE
1 Amber 67
₹17,600.00INR / Case of 144
DWK Life Sciences (Kimble) Score-Break Ampule, Amber, Trimmed Stem, Gross Pack, 2 mL; 144/CS 3459507 Mfr # 12040G-2
COMPARE
2 Amber 75
₹26,400.00INR / Case of 144
DWK Life Sciences (Kimble) Score-Break Ampule, Amber, Trimmed Stem, Large Pack, 20 mL; 1152/CS 3459510 Mfr # 12040U-20
COMPARE
20 Amber 135
₹227,000.00INR / Case of 1152
DWK Life Sciences (Kimble) Score-Break Ampule, Clear, Short Stem, Gross Pack, 2 mL; 144/CS 3459512 Mfr # 12050G-2
COMPARE
2 Clear 61
₹17,200.00INR / Case of 144