Drum and Tank Samplers for Viscous Substances
Draw up viscous media with powerful suction
  • Poreless surfaces are easy to clean
Simple to use—just pull on the T-shaped handle to collect medium-viscosity materials such as slurries, wet clay, oils, emulsions, and creams. Available in economic polypropylene (PP) for general sample collection and chemically inert PTFE/FEP to ensure contamination-free sampling.
142,000.00 - ₹312,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Burkle 5331-1060 Sampling Tube for Viscosity Substances, PP, 160 mL 0604177 Mfr # 5331-1060
COMPARE
Sampling Tube for Viscosity Substances, 2 ft depth
₹142,000.00INR / Each
Burkle 5331-1200 Sampling Tube for Viscosity Substances, PP, 650 mL 0604179 Mfr # 5331-1200
COMPARE
Sampling Tube for Viscosity Substances, 6.6 ft depth
₹249,000.00INR / Each
Burkle 5331-2060 Sampling Tube for Viscosity Substances, PTFE/FEP, 160 mL 0604180 Mfr # 5331-2060
COMPARE
Sampling Tube for Viscosity Substances, 2 ft depth
₹194,000.00INR / Each
Burkle 5331-2100 Sampling Tube for Viscosity Substances, PTFE/FEP, 300 mL 0604181 Mfr # 5331-2100
COMPARE
Sampling Tube for Viscosity Substances, 3.3 ft depth
₹221,000.00INR / Each
Burkle 5331-2200 Sampling Tube for Viscosity Substances, PTFE/FEP, 650 mL 0604182 Mfr # 5331-2200
COMPARE
Sampling Tube for Viscosity Substances, 6.6 ft depth
₹312,000.00INR / Each