HF Scientific DPD Powder Pop Dispensers
Convenient no spill DPD powder dispensing
  • Precise dosing with simple push button operation
  • No direct powder or foil packet handling
Deliver the correct volume of DPD reagent for free or total chlorine accurately using these portable dispensers. Simply invert the dispenser over the sample tube and press the button to dispense reagent. The powder dissolves quickly in the sample with no messy residue.
Use the DPD powder formulation for Hach, HF scientific, and Hanna colorimeters. Use the tablet formulation for LaMotte, Orbeco-Hellige, Palintest, Thermo Scientific Orion, and Tintometer units.
Dispensers provide enough DPD powder for 100 tests for 5-mL samples or order multi-packs of five dispensers with 200 tests each for 10-mL or 25-mL samples.
11,300.00 - ₹109,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
HF Scientific 10500 Dispenser for Free Chlorine, 100 Tests, 5 Ml 9956011 Mfr # 10500
COMPARE
Dispenser for Free Chlorine, 100 Tests, 5 mL
₹11,300.00INR / Each
HF Scientific 10500C Dispenser for Free Chlorine, 1000 Tests, 5 mL 9956012 Mfr # 10500C
COMPARE
Dispenser for Free Chlorine, 1000 Tests, 5 mL
₹93,600.00INR / Pkg of 1000
HF Scientific 10502 Dispenser for Total Chlorine, 100 Tests, 5 Ml 9956013 Mfr # 10502
COMPARE
Dispenser for Total Chlorine, 100 Tests, 5 mL
₹11,300.00INR / Each
HF Scientific 10502C Dispenser for Total Chlorine, 1000 Tests, 5 Ml 9956014 Mfr # 10502C
COMPARE
Dispenser for Total Chlorine, 1000 Tests, 5 mL
₹93,600.00INR / Pkg of 1000
HF Scientific 10504 Dispenser for Free Chlorine, 100 Tests, 10 Ml 9956015 Mfr # 10504
COMPARE
Dispenser for Free Chlorine, 100 Tests, 10 mL
₹14,200.00INR / Each
HF Scientific 10504C Dispenser for Free Chlorine, 1000 Tests, 10 Ml 9956016 Mfr # 10504C
COMPARE
Dispenser for Free Chlorine, 1000 Tests, 10 mL
₹109,000.00INR / Pkg of 1000
HF Scientific 10506 Dispenser for Total Chlorine, 100 Tests, 10 Ml 9956017 Mfr # 10506
COMPARE
Dispenser for Total Chlorine, 100 Tests, 10 mL
₹14,200.00INR / Each
HF Scientific 10506C Dispenser for Total Chlorine, 1000 Tests, 10 Ml, 5 dispensers 9956018 Mfr # 10506C
COMPARE
Dispenser for Total Chlorine, 1000 Tests, 10 mL
₹109,000.00INR / Pkg of 1
HF Scientific 10504T Dispenser, Free Chlorine, Tablet form, 10 Ml; 100/Pk 9956019 Mfr # 10504T
COMPARE
Dispenser for Free Chlorine, 100 Tests, 10 mL
₹14,200.00INR / Each
HF Scientific 10505T Dispenser, Free Chlorine, Tablet form, 10 Ml; 1000/Pk 9956020 Mfr # 10505T
COMPARE
Dispenser for Free Chlorine, 1000 Tests, 10 mL
₹109,000.00INR / Pkg of 1
HF Scientific 10506T Dispenser, Total Chlorine, Tablet form, 10 Ml; 100/Pk 9956021 Mfr # 10506T
COMPARE
Dispenser for Total Chlorine, 100 Tests, 10 mL
₹14,200.00INR / Each
Showing 1- 11 of 11