HF Scientific 10504C Dispenser for Free Chlorine, 1000 Tests, 10 Ml
Representative image only.
HF Scientific 10504C Dispenser for Free Chlorine, 1000 Tests, 10 Ml
HF Scientific | Mfr: 10504C| Item: 9956016
Convenient no spill DPD powder dispensing
₹109,000.00INR / Pkg of 1000
₹109,000.00INR / Pkg of 1000
Product Description
Key Features
  • Precise dosing with simple push button operation
  • No direct powder or foil packet handling
More About this Item
Deliver the correct volume of DPD reagent for free or total chlorine accurately using these portable dispensers. Simply invert the dispenser over the sample tube and press the button to dispense reagent. The powder dissolves quickly in the sample with no messy residue.
Use the DPD powder formulation for Hach, HF scientific, and Hanna colorimeters. Use the tablet formulation for LaMotte, Orbeco-Hellige, Palintest, Thermo Scientific Orion, and Tintometer units.
Dispensers provide enough DPD powder for 100 tests for 5-mL samples or order multi-packs of five dispensers with 200 tests each for 10-mL or 25-mL samples.
Specifications
  • Detector TypePowder formulation for Hach, HF scientific, and Hanna meters
  • Test ParametersFree Chlorine
  • Number of Tests Included1000
  • DescriptionDispenser for Free Chlorine, 1000 Tests, 10 mL