Dilvac Shallow Form Stainless Steel Dewar Flask
Save on coolant
The wide-mouth, shallow dish-shaped glass Dewars, protected by a stainless steel exterior, are ideal for rapid freezing tempering of stirring rods and pestles while requiring only a minimum amount of coolant. The flasks can be used with magnetic stirrers and standard boiling flasks up to 2 L in capacity.
43,800.00 - ₹63,500.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (Liters)
Availability Pricing
COMPARE
0.17
₹43,800.00INR / Each
COMPARE
0.38
₹49,800.00INR / Each
COMPARE
0.57
₹50,700.00INR / Each
COMPARE
1
₹52,400.00INR / Each
COMPARE
1.9
₹63,500.00INR / Each