Digi-Sense Temperature Probe Thermocouple Extension Cables - Standard
Are 20-gauge (0.032" dia) Thermocouple Wire with PVC/PVC Insulation
  • Maximum temperature: 221° F (105° C).
  • Cables come in a variety of lengths.
  • Each probe comes with color coded ANSI standard connectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
4,300.00 - ₹10,900.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Length (in) Use With Probe / Wire Type
Availability Pricing
COMPARE
Standard Connector
₹4,300.00INR / Each
COMPARE
Standard Connector
₹10,900.00INR / Each
COMPARE
Standard Connector
₹4,500.00INR / Each
COMPARE
Standard Connector
₹7,500.00INR / Each
COMPARE
Standard Connector
₹10,900.00INR / Each
COMPARE
Standard Connector
₹4,600.00INR / Each
COMPARE
Standard Connector
₹7,500.00INR / Each
Showing 1- 7 of 7