Digi-Sense Blue Spirit PFA Coated Glass Thermometers, Fahrenheit-scale
The liquid and glass are contained within the PTFE jacket, preventing contamination
  • Nonmercury blue spirit liquid
  • Statement of accuracy
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage, the PTFE coating contains the broken liquid and glass, preventing contamination or injury.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
COMPARE
230 0 300
Call For Price
COMPARE
300 0 300
Call For Price
COMPARE
230 0 300