Digi-Sense® Blue Spirit ASTM Glass Thermometers
Pre-calibrated to save you time
  • Mercury-free thermometers conform to ASTM specifications
  • Individually serialized for easy identification
  • Blue organic fill is easy to see against backdrop of white glass
These NIST-traceable thermometers fulfill requirements for performing ASTM methods without the worry of mercury contamination. Organic fill solution is safer to clean up in the event of breakage. The calibration report states corrections to one-tenth of the smallest division, providing an accurate offset for ensuring precise measurement records.
73,200.00 - ₹80,200.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
ASTM Number Description
Availability Pricing
COMPARE
S62C-03 ASTM S62C-03 Individually Calibrated Liquid-In-Glass Thermometer; -38/2C, Organic Liquid Fill
₹80,200.00INR / Each
COMPARE
S63C-03 ASTM S63C-03 Individually Calibrated Liquid-In-Glass Thermometer; -8/32C, Organic Liquid Fill
₹73,200.00INR / Each
COMPARE
S63F-03 ASTM S63F-03 Individually Calibrated Liquid-In-Glass Thermometer; 18/89F, Organic Liquid Fill
₹73,200.00INR / Each
COMPARE
S64C-03 ASTM S64C-03 Individually Calibrated Liquid-In-Glass Thermometer; 25/55C, Organic Liquid Fill
₹80,200.00INR / Each
COMPARE
S64F-03 ASTM S64F-03 Individually Calibrated Liquid-In-Glass Thermometer; 77/131F, Organic Liquid Fill
₹80,200.00INR / Each
COMPARE
S65C-03 ASTM S65C-03 Individually Calibrated Liquid-In-Glass Thermometer; 50/80C, Organic Liquid Fill
₹80,200.00INR / Each
COMPARE
S66C-03 ASTM S66C-03 Individually Calibrated Liquid-In-Glass Thermometer; 75/105C, Organic Liquid Fill
₹80,200.00INR / Each
COMPARE
S67C-03 ASTM S67C-03 Individually Calibrated Liquid-In-Glass Thermometer; 95/155C, Organic Liquid Fill
₹80,200.00INR / Each
Showing 1- 8 of 8