Diba Omnifit® PTFE Cap System Union Fittings and Valves
Create connections with very low dead volume
  • Select fittings with on/off valve to control your fluid line
  • Use with 1 to 4 mm OD tubing
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.
18,700.00 - ₹58,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (mm) Fitting Material Orifice Dimensions (mm)
Availability Pricing
Diba Omnifit® Reducing Connector, 4 to 11 x 1 to 4 mm OD tubing, 2-way, straight; 2/pk 2193907 Mfr # 001008
COMPARE
11 PTFE/PP 1.5
₹25,200.00INR / Pkg of 2
Diba Omnifit® Standard Connector for 1 to 4 mm OD Tubing, 2-way, elbow; 1/ea 2193909 Mfr # 01011
COMPARE
4 PTFE/PP 1.5
₹18,700.00INR / Each
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 2-way, straight, mountable; 2/pk 2193916 Mfr # 1104
COMPARE
4 PTFE/PP 1.5
₹58,300.00INR / Pkg of 2