Diba Omnifit® PTFE Cap System Straight Fittings and Valves
Connect various types of tubing with low dead volume
  • Select fittings with on/off valve to control your fluid lines
  • Use with 1 to 4 mm OD tubing
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.
11,800.00 - ₹29,100.00INR / Pkg of 3
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Valve Material Orifice Dimensions (mm)
Availability Pricing
Diba Omnifit® Standard Connector for 1 to 4 mm OD Tubing, 2-way, straight; 3/pk WW-21939-00 Mfr # 001001
COMPARE
PTFE 1.5
₹11,800.00INR / Pkg of 3
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 2-way, straight; 2/pk WW-21939-13 Mfr # 001101
COMPARE
PTFE 1.5
₹29,100.00INR / Pkg of 2