Diba Omnifit® Flared Tubing Assemblies with Click-N-Seal® 1/4"-28 UNF(M) Fittings, 1/16"ID x 1/8"OD
Flared tubing and fitting assemblies offer fast leak-free connections
  • When the cap clicks, you know the seal is tight
  • Flared tubing sits tightly against a flat bottom of the port
  • Only the tube material is in the fluid path
  • Available with polycarbonate (PC), acetal, and PEEK fittings
Self-torquing Click-N-Seal fittings create a seal without tools. As the fitting is tightened the resistance of the cap increases until the correct torque is reached. Acetal and PEEK fittings' compact design easily fits in tight spaces.
4,900.00 - ₹5,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing Formulation
Availability Pricing
Diba Click-N-Seal™ Plus Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/Acetal, 1/8" OD x 24" L; 1/ea 2194347 Mfr # DB200-15050
COMPARE
2 PTFE
₹5,000.00INR / Each
Diba Click-N-Seal™ Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/PC, 1/8" OD x 24" L; 1/ea 2194350 Mfr # DB200-15053
COMPARE
2 PTFE
₹4,900.00INR / Each