Diba Dibafit™ Type S Ferrules for Click-N-Seal Fittings with Micro 6-40 UNF(M) Thread
Fitting and ferrule solution for quick and easy connection of 1/16" tube into a 6-40 threaded port
  • Pressure rating up to 500 psi (34 bar)
  • 316 stainless steel case with PEEK™ ferrule insert and 6-40 thread fitting
  • Retain 1/16" OD PTFE tube with 0.3, 0.5 or 0.8mm bore
This fitting system uses a two-piece ferrule which fits permanently to the outside of the tubing. Two-part ferrules consist of an inner PEEK™ sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path. The fitting can spin freely, eliminating tubing twist. A recess in the fitting houses the ferrule, allowing maximum thread engagement with the port.
10,500.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Diba Omni-Lok™ Micro Type-S Ferrule, 1/16" OD tubing, PEEK™ / SS, taper, 0.3 mm bore; 10/pk WW-21940-50 Mfr # 008FK16-03C9L
COMPARE
PEEK/SS Omni-Lok™ Micro Type-S Ferrule, 1/16\" OD tubing, PEEK / SS, taper, 0.3 mm bore; 10/pk
₹10,500.00INR / Pkg of 10
Diba Omni-Lok™ Micro Type-S Ferrule, 1/16" OD tubing, PEEK™ / SS, taper, 0.5 mm bore; 10/pk WW-21940-51 Mfr # 008FK16-05C9L
COMPARE
PEEK/SS Omni-Lok™ Micro Type-S Ferrule, 1/16\" OD tubing, PEEK / SS, taper, 0.5 mm bore; 10/pk
₹10,500.00INR / Pkg of 10
Diba Omni-Lok™ Micro Type-S Ferrule, 1/16" OD tubing, PEEK™ / SS, taper, 0.8 mm bore; 10/pk WW-21940-52 Mfr # 008FK16-08C9L
COMPARE
PEEK/SS Omni-Lok™ Micro Type-S Ferrule, 1/16\" OD tubing, PEEK / SS, taper, 0.8 mm bore; 10/pk
₹10,500.00INR / Pkg of 10
Diba Omni-Lok™ Micro Type-S Ferrule, 1/16" OD tubing, PEEK™ / SS, flat bottom, 0.8 mm bore; 10/pk WW-21940-54 Mfr # 008FK16-F9L
COMPARE
PEEK/SS Omni-Lok™ Micro Type-S Ferrule, 1/16\" OD tubing, PEEK / SS, flat bottom, 0.8 mm bore; 10/pk
₹10,500.00INR / Pkg of 10