Diba Click-N-Seal® Fittings with M6 ISO(M) Thread for Flared Tubing
Achieve a leak-free seal with only the tube material in the fluid path
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
Fittings for flared tubing create a ferrule-free connection and ensure leak-free seal while minimizing carryover. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. A seal is created by compressing the flared portion of the tube against the flat bottom of the port.
Select from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance. Choose the size that best matches your tube OD. A washer is recommended to ensure the flare seals correctly. Use flanging kit to easily flange semi-rigid tubing.
3,800.00 - ₹7,900.00INR / Pkg of 6
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Diba Click-N-Seal™ Plus Tubing End Fitting, 1/16" OD flared tubing, M6 ISO(M), white acetal; 6/pk WW-21942-41 Mfr # 008NG16-AT6H
COMPARE
White 1/16 Acetal
₹7,900.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, M6 ISO(M), black PC; 6/pk WW-21942-45 Mfr # 008NG16-CS6B
COMPARE
Black 1/16 Polycarbonate
₹3,800.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, M6 ISO(M), white PC; 6/pk WW-21942-46 Mfr # 008NG16-CS6H
COMPARE
White 1/16 Polycarbonate
₹3,800.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/16" OD flared tubing, M6 ISO(M), red PC; 6/pk WW-21942-47 Mfr # 008NG16-CS6R
COMPARE
Red 1/16 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Plus Tubing End Fitting, 1.6 to 2.5 mm OD flared tubing, M6 ISO(M), acetal; 6/pk WW-21942-50 Mfr # 008NG25-AT6H
COMPARE
White 3/32 Acetal
₹7,900.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5 mm OD flared tubing, M6 ISO(M), black PC; 6/pk WW-21942-54 Mfr # 008NG25-CS6B
COMPARE
Black 3/32 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5 mm OD flared tubing, M6 ISO(M), white PC; 6/pk WW-21942-55 Mfr # 008NG25-CS6H
COMPARE
White 3/32 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1.6 to 2.5 mm OD flared tubing, M6 ISO(M), red PC; 6/pk WW-21942-56 Mfr # 008NG25-CS6R
COMPARE
Red 3/32 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, M6 ISO(M), white acetal; 6/pk WW-21942-59 Mfr # 008NG32-AT6H
COMPARE
White 1/8 Acetal
₹7,900.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, M6 ISO(M), black PC; 6/pk WW-21942-63 Mfr # 008NG32-CS6B
COMPARE
Black 1/8 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, M6 ISO(M), white PC; 6/pk WW-21942-64 Mfr # 008NG32-CS6H
COMPARE
White 1/8 Polycarbonate
₹5,200.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, 1/8" OD flared tubing, M6 ISO(M), red PC; 6/pk WW-21942-65 Mfr # 008NG32-CS6R
COMPARE
Red 1/8 Polycarbonate
₹5,200.00INR / Pkg of 6
Showing 1- 12 of 12