Diba Click-N-Seal® Fittings with M6 ISO(M) Thread for Dibafit™ Inverted Cone System
Removable and reusable system for quick and convenient low-pressure connection
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
Use fittings with inverted cones to achieve a simple and easy-to-use low-pressure connection. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. The ETFE cone is the only part in the fluid path and offers excellent chemical resistance. The cone sits in the recessed area of the fitting allowing maximum thread engagement. None of the parts are permanently attached to the tubing, so both fittings and inverted cones can easily be removed and reused. Select from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
3,800.00 - ₹12,100.00INR / Pkg of 6
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Diba Click-N-Seal™ Plus Tubing End Fitting, inverted cone, 1/16" OD tubing, M6 ISO(M), acetal; 6/pk WW-21941-61 Mfr # 008NC16-AT6H
COMPARE
White 1/16 Acetal
₹6,000.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16" OD tubing, M6 ISO(M), black PC; 10/pk WW-21941-65 Mfr # 008NC16-CS6B
COMPARE
Black 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16" OD tubing, M6 ISO(M), white PC; 10/pk WW-21941-66 Mfr # 008NC16-CS6H
COMPARE
White 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16" OD tubing, M6 ISO(M), red PC; 10/pk WW-21941-67 Mfr # 008NC16-CS6R
COMPARE
Red 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Tubing End Fitting, inverted cone, 1/16" OD tubing, M6 ISO(M), PEEK; 10/pk WW-21941-69 Mfr # 008NC16-KT6L
COMPARE
Natural 1/16 PEEK
₹12,100.00INR / Pkg of 10
Diba Click-N-Seal™ Plus Tubing End Fitting, inverted cone, 1/8" OD tubing, M6 ISO(M), acetal; 10/pk WW-21941-76 Mfr # 008NC32-AT6H
COMPARE
White 1/8 Acetal
₹7,900.00INR / Pkg of 1
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/8" OD tubing, M6 ISO(M), black PC; 10/pk WW-21941-80 Mfr # 008NC32-CS6B
COMPARE
Black 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/8" OD tubing, M6 ISO(M), white PC; 10/pk WW-21941-81 Mfr # 008NC32-CS6H
COMPARE
White 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/8" OD tubing, M6 ISO(M), red PC; 10/pk WW-21941-82 Mfr # 008NC32-CS6R
COMPARE
Red 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Tubing End Fitting, inverted cone, 1/8" OD tubing, M6 ISO(M), PEEK; 10/pk WW-21941-84 Mfr # 008NC32-KT6L
COMPARE
Natural 1/8 PEEK
₹12,100.00INR / Pkg of 10
Showing 1- 10 of 10