Diba Click-N-Seal® Fittings with Flat-Bottom 1/4"-28 UNF(M) Thread for Dibafit™ Type P/S Ferrule System
Permanently attached, chemically inert ferrule is ideal for repeated connect/disconnect
  • Excellent for high-pressure application
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
Permanently attached ferrule allow fittings for use in high-pressure applications. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. The fitting can spin freely from tubing, preventing tubing from twisting. A recess in the fitting houses the ferrule, allowing maximum thread engagement with the port. Select fittings from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
Both Type P and Type S ferrules fits permanently onto the outside of semi-rigid tubing. Type P system uses a one-piece ferrule with only the inner PTFE sealing face in the fluid path. Type S system uses a two-piece ferrule made up of an inner PEEK or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
3,800.00 - ₹12,100.00INR / Pkg of 10
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 1/4"-28 UNF(M), black PC; 10/pk WW-21941-94 Mfr # 008NF16-CS5B
COMPARE
Black 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 1/4"-28 UNF(M), white PC; 10/pk WW-21941-95 Mfr # 008NF16-CS5H
COMPARE
White 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/16" OD tubing, 1/4"-28 UNF(M), red PC; 10/pk WW-21941-96 Mfr # 008NF16-CS5R
COMPARE
Red 1/16 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Tubing End Fitting, Type S/P, 1/16" OD tubing, 1/4"-28 UNF(M), PEEK; 10/pk WW-21942-02 Mfr # 008NF16-KT5L
COMPARE
Natural 1/16 PEEK
₹12,100.00INR / Pkg of 10
Diba Click-N-Seal™ Plus Tubing End Fitting, Type S/P, 1/8" OD tubing, 1/4"-28 UNF(M), acetal; 6/pk WW-21942-18 Mfr # 008NF32-AT5H
COMPARE
White 1/8 Acetal
₹6,000.00INR / Pkg of 6
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, 1/4"-28 UNF(M), black PC; 10/pk WW-21942-20 Mfr # 008NF32-CS5B
COMPARE
Black 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, 1/4"-28 UNF(M), white PC; 10/pk WW-21942-21 Mfr # 008NF32-CS5H
COMPARE
White 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Tubing End Fitting, Type S/P, 1/8" OD tubing, 1/4"-28 UNF(M), red PC; 10/pk WW-21942-22 Mfr # 008NF32-CS5R
COMPARE
Red 1/8 Polycarbonate
₹3,800.00INR / Pkg of 10
Diba Click-N-Seal™ Ultra Tubing End Fitting, Type S/P, 1/8" OD tubing, 1/4"-28 UNF(M), PEEK; 10/pk WW-21942-26 Mfr # 008NF32-KT5L
COMPARE
Natural 1/8 PEEK
₹12,100.00INR / Pkg of 10
Showing 1- 9 of 9