Delmhorst® QuickNav Moisture Meter
Get quick and accurate temperature and RH readings
  • Hold feature allows you to hold the displayed reading when testing hard-to-reach areas
  • Backlit LCD screen makes reading in dark places easy
  • Alarm alerts when preselected moisture level is reached
  • Thermohygrometer conforms to ASTM F-2170 standard for concrete testing
Versatile 3-in-1 handheld moisture meter offers pin mode, scan mode, and thermohygrometer functions in a single easy-to-use unit. Meter quickly and accurately measures moisture, temperature, and RH, and also calculates dew point, absolute humidty, and vapor pressure. Pin penetration depth is 5/16” and scan depth is ¾”.
218,000.00 - ₹266,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Type Moisture Range Wood Scale (Max) (%) Description
Availability Pricing
Delmhorst QUICKNAV/BP Moisture Meter Basic Kit 5982327 Mfr # QUICKNAV/BP
COMPARE
Pin 60 Moisture Meter Basic Kit
₹218,000.00INR / Each
Delmhorst QUICKNAV/CP Moisture Meter Kit 5982328 Mfr # QUICKNAV/CP
COMPARE
Pin 60 Moisture Meter Kit
₹266,000.00INR / Each