Delmhorst® PX-30 Paper Moisture Meters
Combines the advanced design of the PX-20 with added moisture scales for baled paper and a reference scale, making it an essential tool for the paper industry to ensure consistent quality and avoid moisture issues
  • Moisture ranges for kraft paper (4.3-18%), baled scrap paper (5-40%), and relative readings for other materials
  • Custom display with auto-backlight and on-screen stats for up to 100 readings
  • Bluetooth® connectivity to the EDGE® app for data export, timestamping, and geotagging
  • Color-coded LED alerts, customizable through the EDGE® app, and alarm set points for precision monitoring
  • Durable ABS case with E-Z grip handle and ergonomic design for easy handling
The PX-30 Paper Moisture Meter is trusted in the paper industry for its accuracy, ease of use, and versatility. Designed for both kraft and baled scrap paper, it also offers a reference scale for moisture readings in other materials. The PX-30 connects to the Delmhorst® EDGE® app, enabling users to export data, customize LED ranges, and tag readings for thorough documentation. With a rugged, ergonomic case and intuitive display, the PX-30 is perfect for ensuring paper quality and consistency on every jobsite.
118,000.00 - ₹146,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type Measurement Type Description
Availability Pricing
Delmhorst PX-30 Paper Moisture Meter with 324CAS-0112 Case 5980089 Mfr # PX-30/P01
COMPARE
None included Paper (4.3-18%), Baled Scrap Paper (5-40%), 0-100 Reference Scale Paper Moisture Meter with 324CAS-0112 Case
₹120,000.00INR / Each
Delmhorst PX-30 Paper Moisture Meter with 324CAS-0112 Case, H-4 Handle, 830-2 10" Prod 5980090 Mfr # PX-30/P02
COMPARE
830-2 10" prod Paper (4.3-18%), Baled Scrap Paper (5-40%), 0-100 Reference Scale Paper Moisture Meter with 324CAS-0112 Case, H-4 Handle, 830-2 10" Prod
₹146,000.00INR / Each
Delmhorst PX-30 Paper Moisture Meter with 324CAS-0111 Case 5980091 Mfr # PX-30W/CS
COMPARE
None included Paper (4.3-18%), Baled Scrap Paper (5-40%), 0-100 Reference Scale Paper Moisture Meter with 324CAS-0111 Case
₹118,000.00INR / Each